Learn More
Mopec™ Waffle Specimen Chucks for Leica, TBS, and Tanner Cryostats
Waffle Specimen Chucks are available in multiple sizes and colors.
Supplier:  Mopec™ SP103WPR
 View more versions of this product
Catalog No. 22-445-450
Only null left
Explore available promotions
Description
Description
  • Round, aluminum specimen chucks
  • Suitable for Leica (except CM1950 model), TBS, and Tanner Cryostats
Specifications
Specifications
Leica, TBS, and Tanner Cryostats
30 mm
Purple
Product Suggestions
Product Suggestions