missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mopec™ Waffle Specimen Chucks for Leica, TBS, and Tanner Cryostats
Description
- Round, aluminum specimen chucks
- Suitable for Leica (except CM1950 model), TBS, and Tanner Cryostats
Specifications
Specifications
| For Use With (Equipment) | Leica, TBS, and Tanner Cryostats |
| Diameter (Metric) | 30 mm |
| Color | Blue |