missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Sulfate, Heptahydrate, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
Description
Spectrum™ Chemical Ferrous Sulfate, Heptahydrate, Crystal, Reagent, ACS As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Specifications
Specifications
| Physical Form | Crystals or Needles |
| CAS Max % | 100% |
| Quantity | 50 kg |
| Packaging | Amber Glass Bottle |
| Percent Purity | 99% |
| Grade | ACS/Reagent |
| Chemical Name or Material | Ferrous Sulfate, Heptahydrate, Crystal |