missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VPS39 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521104
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521104 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521104 Supplier Invitrogen™ Supplier No. PA521104
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

A suggested positive control is rat liver tissue lysate. PA5-21104 can be used with blocking peptide PEP-1218.

VPS39 is the mammalian homolog of the yeast vacuolar protein VAM6, that in mammals, promotes lysosome clustering and fusion in vivo. VPS39 is also thought to modulate the TGF-beta response by coupling the TGF-beta receptor complex to the Smad pathway. Similar to the related protein TRAP1, the VPS39 protein is essential for early embryonic development, with mice lacking VPS39 dying at or before E6. 5.

Specifications

Antigen VPS39
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene VPS39
Gene Accession No. Q8R5L3, Q96JC1
Gene Alias A230065P22Rik; AW743070; hVam6p; KIAA0770; mVam6; OTTMUSP00000023341; OTTMUSP00000023350; PI3K type 3; Pldn; TLP; TRAP1-like protein; vacuolar protein sorting 39 (yeast); vacuolar protein sorting 39 homolog; vacuolar protein sorting 39 homolog (S. cerevisiae); Vam6; Vam6/Vps39-like protein; Vam6P; Vps39; VPS39 HOPS complex subunit; VPS39, HOPS complex subunit
Gene Symbols VPS39
Host Species Rabbit
Immunogen An 18 amino acid peptide near the carboxy terminus of human VPS39.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23339, 269338, 362199
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less