missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS37A Polyclonal Antibody

Description
Antibody detects endogenous levels of total VPS37A.
This gene belongs to the VPS37 family, and encodes a component of the ESCRT-I (endosomal sorting complex required for transport I) protein complex, required for the sorting of ubiquitinated transmembrane proteins into internal vesicles of multivesicular bodies. Expression of this gene is downregulated in hepatocellular carcinoma, and mutations in this gene are associated with autosomal recessive spastic paraplegia-53. A related pseudogene has been identified on chromosome 5. Alternatively spliced transcript variants have been found for this gene.
Specifications
Specifications
| Antigen | VPS37A |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Vps37a |
| Gene Accession No. | Q8CHS8, Q8NEZ2 |
| Gene Alias | 2210018P21Rik; 4930592A21Rik; AW261445; D8Ertd531e; ESCRT-I complex subunit VPS37A; HCRP1; Hepatocellular carcinoma-related protein 1; hVps37A; polyglutamine binding protein 2; PQBP2; SPG53; vacuolar protein sorting 37 homolog A; vacuolar protein sorting 37 homolog A (S. cerevisiae); vacuolar protein sorting 37A; vacuolar protein sorting 37A (yeast); vacuolar protein sorting-associated protein 37A; Vps37A; VPS37A, ESCRT-I subunit |
| Gene Symbols | Vps37a |
| Show More |