missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS35 Recombinant Rabbit Monoclonal Antibody (23GB4260)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553721
Description
VPS35 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex.Specifications
| VPS35 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| VPS35 | |
| AI647796; DKFZp434E1211; DKFZp434P1672; FLJ10752; FLJ13588; FLJ20388; hVPS35; Maternal embryonic message 3; maternal-embryonic 3; Mem3; PARK17; TCCCTA00141; vacuolar protein sorting 35; vacuolar protein sorting 35 homolog; vacuolar protein sorting 35 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 35; Vesicle protein sorting 35; VPS35; VPS35 retromer complex component | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25479, 55737, 65114 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4260 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96QK1, Q9EQH3 | |
| VPS35 | |
| A synthesized peptide derived from VPS35 (150-200AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |