Learn More
Invitrogen™ VorTemp™ 56 Shaking Incubator and Temperature Validation Kit
The VorTemp 56 microplate shaking incubator is designed for simultaneous heating and mixing of QuantiGene Plex assays. The temperature is adjustable over a broad range, and mechanical convection provides a uniform and stable environment within the chamber.
$5,900.00
Specifications
| Temperature Range (Metric) | 4°C to 99.5°C |
|---|---|
| Product Type | Shaking Incubator |
| Material | Coated Steel |
| Humidity Range | 85% |
| Includes | 1 VorTemp 56 Shaking Incubator, 1 QuantiGene Temperature Validation Kit |
Description
The VorTemp 56 microplate shaking incubator is designed for simultaneous heating and mixing of QuantiGene Plex assays. The temperature is adjustable over a broad range, and mechanical convection provides a uniform and stable environment within the chamber.
VorTemp features:
- A microprocessor to control all parameters including temperature and timed operation
- Self-diagnostic software to continuously monitor for proper temperature and shaking speed, alerting the user to any errors
- Heating and shaking that are controlled by feedback loops which precisely maintain chosen parameters, independent of load or voltage fluctuations
- Sturdy construction for stability during high speed shaking
- An insulated chamber to maintain temperature and reduce operating noise
Specifications
| 4°C to 99.5°C | |
| Coated Steel | |
| 1 VorTemp 56 Shaking Incubator, 1 QuantiGene Temperature Validation Kit |
| Shaking Incubator | |
| 85% | |
| 1 Ea. |
For Research Use Only. Not for use in diagnostic procedures.