missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VorTemp™ 56 Shaking Incubator and Temperature Validation Kit

Catalog No. QP0703
Change view
Click to view available options
Voltage:
120 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
QP0703 120 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QP0703 Supplier Invitrogen™ Supplier No. QP0703

The VorTemp 56 microplate shaking incubator is designed for simultaneous heating and mixing of QuantiGene Plex assays. The temperature is adjustable over a broad range, and mechanical convection provides a uniform and stable environment within the chamber.

The VorTemp 56 microplate shaking incubator is designed for simultaneous heating and mixing of QuantiGene Plex assays. The temperature is adjustable over a broad range, and mechanical convection provides a uniform and stable environment within the chamber.

VorTemp features:

  • A microprocessor to control all parameters including temperature and timed operation
  • Self-diagnostic software to continuously monitor for proper temperature and shaking speed, alerting the user to any errors
  • Heating and shaking that are controlled by feedback loops which precisely maintain chosen parameters, independent of load or voltage fluctuations
  • Sturdy construction for stability during high speed shaking
  • An insulated chamber to maintain temperature and reduce operating noise

Specifications

Voltage 120 V
Temperature Range (Metric) 4°C to 99.5°C
Product Type Shaking Incubator
Material Coated Steel
Humidity Range 85%
Includes 1 VorTemp 56 Shaking Incubator, 1 QuantiGene Temperature Validation Kit
Quantity 1 Ea.

For Research Use Only. Not for use in diagnostic procedures.