missing translation for 'onlineSavingsMsg'
Learn More

Voriconazole 98.0+%, TCI America™

Catalog No. p-7137930
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0116100MG 100 mg
V01161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0116100MG Supplier TCI America Supplier No. V0116100MG

Chemical Identifiers

CAS 137234-62-9
Molecular Formula C16H14F3N5O
Molecular Weight (g/mol) 349.317
MDL Number MFCD00905717
InChI Key BCEHBSKCWLPMDN-MGPLVRAMSA-N
Synonym voriconazole, vfend, 2r,3s-2-2,4-difluorophenyl-3-5-fluoropyrimidin-4-yl-1-1h-1,2,4-triazol-1-yl butan-2-ol, voriconazol, voriconazolum, voriconazole usan:inn:ban, unii-jfu09i87tr, vfend tn, 2r,3s-2-2,4-difluorophenyl-3-5-fluoropyrimidin-4-yl-1-1,2,4-triazol-1-yl butan-2-ol, vcz
PubChem CID 71616
ChEBI CHEBI:10023
IUPAC Name (2R,3S)-2-(2,4-difluorophenyl)-3-(5-fluoropyrimidin-4-yl)-1-(1,2,4-triazol-1-yl)butan-2-ol
SMILES CC(C1=NC=NC=C1F)C(CN2C=NC=N2)(C3=C(C=C(C=C3)F)F)O

Specifications

Melting Point 132°C
Color White
Quantity 100 mg
UN Number 2811
Formula Weight 349.32
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Voriconazole