missing translation for 'onlineSavingsMsg'
Learn More

Voriconazole Pharmaceutical Secondary Standard:, MilliporeSigma™ Supelco™

Catalog No. 111018538
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018538 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018538 Supplier Merck Emd Millipore Supplier No. PHR18921G
Voriconazole is a synthetically prepared, broad-spectrum triazole derivative of fluconazole, which shows in vitro activity against many yeasts and a broad-spectrum of mold and dermatophyte isolates. Its mode of action involves the inhibition of cytochrome P450 (CYP)-dependent enzyme, 14-α-sterol demethylase, and hence it is involved in disrupting the cell membrane and terminate the fungal growth.

Specifications

CAS 137234-62-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C16H14F3N5 O
MDL Number MFCD00905717
Synonym Voriconazole; 2 R,3 S-2-(2,4-Difluorophenyl)-3-(5-fluoropyrimidin-4-yl)-1-(1 H-1,2,4-triazol-1-yl)butan-2-ol; UK-109496
Type Certified Reference Material
Quantity 1 g
Formula Weight 349.31
Grade Certified Reference Material