missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Volociximab Chimeric Recombinant Mouse Monoclonal Antibody (M200 (Volociximab))
GREENER_CHOICE

Catalog No. MA551584
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551584 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551584 Supplier Invitrogen™ Supplier No. MA551584
Only null left

Mouse Recombinant Monoclonal Antibody

The antibody binds to human alpha 5 Beta 1 integrin with a Kd of 0.367 nM (determined by Biacore), an EC50 of 0.2 nM and an IC50 of 2.3 nM.

Volociximab is a biosimilar that targets ITGA5.

Specifications

Antigen Volociximab Chimeric
Applications ELISA, Flow Cytometry, Inhibition Assays, Immunohistochemistry, Immunocytochemistry
Classification Recombinant Monoclonal
Clone M200 (Volociximab)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias 558480-40-3; M20
Host Species Mouse
Immunogen Heterodimeric Human alpha 5 Beta 1 Fc fusion protein.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less