Learn More
Medegen Vollrath Stainless-Steel Beakers without Spout
Supplier:  Medegen 78720
 View more versions of this product
Catalog No. 02-584C
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Aluminum Bonded
Stainless Steel
275°F
Beaker
NSA
1900 mL
Welded Handles
135°C
Sterile
Product Suggestions
Product Suggestions