missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VOAS in Soil - Low Level - PT, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-56-1 |
| Flash Point | 11°C - closed cup - Solvent |
| Type | Proficiency Testing Material |
| Quantity | 30 g |