missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VMP1 Monoclonal Antibody (K92018_1H6), Invitrogen™

Catalog No. MA555722
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555722 50 μL
MA556102 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555722 Supplier Invitrogen™ Supplier No. MA555722
Only null left

Mouse Monoclonal Antibody

VMP1 is a stress-induced protein that, when overexpressed, promotes formation of intracellular vacuoles followed by cell death. May be involved in the cytoplasmic vacuolization of acinar cells during the early stage of acute pancreatitis.VMP1 is a transmembrane protein that co-localizes with LC3, a marker of the autophagosomes. It interacts with beclin-1, a mammalian autophagy initiator, through the VMP1-Atg domain, which is essential for autophagosome formation.

Specifications

Antigen VMP1
Applications Western Blot
Classification Monoclonal
Clone K92018_1H6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene VMP1
Gene Accession No. Q96GC9
Gene Alias 3110098I04Rik; 4930579A11Rik; AI787464; EPG3; HSPC292; MIR21; mir-21a; NF-E2 induceble protein-2; NF-E2-inducible protein 2; ni-2; Protein ni-2; Tango5; TDC1; Tmem49; transmembrane protein 49; Vacuole membrane protein 1; VMP1; wu:fb05d08; wu:fb92g08; zgc:77153
Gene Symbols VMP1
Host Species Mouse
Immunogen Recombinant human VMP1 (immunogen range is 141-250).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81671
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less