missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Vivid™ Regeneration System, 100X
Vivid™ Regeneration System, 100X
Supplier: Invitrogen™ P2878
Description
Vivid™ Regeneration System, 100X, is a Vivid™ CYP450 Screening Kit component consisting of glucose-6-phosphate and glucose-6-phosphate dehydrogenase, which converts NADP+ into NADPH, which is required to start the CYP450 reaction. Vivid™ Regeneration System, 100X, is included in the Vivid™ Screening Kits, but may also be purchased separately.More About Vivid™ CYP450 Screening Kits
Vivid™ CYP450 Screening Kits are high-throughput fluorescence-based assays for detection of enzyme-drug interactions and CYP450 inhibition. Vivid™ CYP450 Screening Kits include Vivid™ Substrate, Vivid™ Fluorescent Standard, reaction buffer, Vivid™ Regeneration System, NADP+, and CYP450 BACULOSOMES™ Plus Reagent. Vivid™ Fluorogenic Substrates are blocked dyes that yield minimal fluorescence signal until cleaved or hydroxylated. Oxidation at either of two potential sites releases the highly fluorescent product. CYP450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells expressing a human CYP450 isozyme and human cytochrome P450 reductase. CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over human liver microsomes in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s.
>> View all Vivid™ CYP450 Screening Kits
For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
Specifications
| VIVID | |
| 1 tube, Store at -80 °C | |
| Regeneration System | |
| Kit | |
| Dry Ice |
| Microplate Reader | |
| Fluorescence | |
| 0.5 mL | |
| Coenzymes |
For Research Use Only. Not for use in diagnostic procedures.