missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ VITLAB™ VITsafe™ Wide-Mouth Wash Bottle

Wide-Mouth Wash Bottle with integrated vent in cap to reduce dripping.

Supplier:  BRANDTECH™ VITLAB™ 1352869

 View more versions of this product

Catalog No. 14380960


Only null left

Description

Description

  • Chemical name in German, English, French and Spanish
  • Chemical formula
  • CAS Number, hazard pictogram, signal word
  • Risk Phrases (H phrases), safety phrases (P phrases)
  • NFPA Code
  • Fine tip polypropylene tube provides directional accuracy
  • Colored cap allows easier identification
Specifications

Chemical Identifiers

500 mL
166 mm
Non-sterile
Reusable
Cylindrical
Safety-labeled Wash Bottle
GL 45
Vented Tube
16.9 oz.
Pack
Yes
No
166 mm
6.54 in.
Spouted Cap
Not Autoclavable

Specifications

500 mL
Orange
LDPE, Polypropylene
Pack
Non-sterile
Yes
No
Cylindrical
Safety-labeled Wash Bottle
6.54 in.
Ethanol
76 mm
Vented Tube
16.9 oz.
Ethanol
166 mm
12
76 mm
Reusable
Wide, Threaded
166 mm
3 in.
6.54 in.
GL 45
Spouted Cap
Not Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions