missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ VITLAB™ VITsafe™ Wide-Mouth Wash Bottle
Wide-Mouth Wash Bottle with integrated vent in cap to reduce dripping.
Supplier: BRANDTECH™ VITLAB™ 1352959
Description
- Chemical name in German, English, French and Spanish
- Chemical formula
- CAS Number, hazard pictogram, signal word
- Risk Phrases (H phrases), safety phrases (P phrases)
- NFPA Code
- Fine tip polypropylene tube provides directional accuracy
- Colored cap allows easier identification
Chemical Identifiers
| 500 mL | |
| 166 mm | |
| Non-sterile | |
| Reusable | |
| Cylindrical | |
| Safety-labeled Wash Bottle | |
| GL 45 | |
| Vented Tube |
| 16.9 oz. | |
| Pack | |
| Yes | |
| No | |
| 166 mm | |
| 6.54 in. | |
| Spouted Cap | |
| Not Autoclavable |
Specifications
| 500 mL | |
| Red | |
| LDPE, Polypropylene | |
| Pack | |
| Non-sterile | |
| Yes | |
| No | |
| Cylindrical | |
| Safety-labeled Wash Bottle | |
| 6.54 in. | |
| GL 45 | |
| Spouted Cap | |
| Not Autoclavable |
| 16.9 oz. | |
| Xylene | |
| 166 mm | |
| 6 | |
| 58 mm | |
| Reusable | |
| Wide | |
| 166 mm | |
| 6.54 in. | |
| Xylene | |
| 76 mm | |
| Vented Tube |