missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Vitronectin (VTN-N) Recombinant Human Protein, Truncated
Description
Vitronectin (VTN-N) is a recombinant human protein that provides a defined surface for feeder-free culture of human pluripotent stem cells (PSCs). When used with Essential 8 Medium (Cat. No. A1517001), VTN-N has been proven to maintain pluripotency and normal growth characteristics in multiple PSC lines.
- Proven to maintain pluripotency and normal growth characteristics in multiple PSC lines when used with Essential 8 Medium
- Supports PSC growth for >50 passages without any signs of karyotypic abnormalities and to maintain the ability of PSCs to differentiate into all three germ line lineages
- Supports human pluripotent stem cell attachment and survival better than wild-type vitronectin when used in conjunction with Essential 8 Medium
- Optimized—designed in the laboratory of James Thomson for use with Essential 8 Medium
- Defined—for reduced variability in your PSC cultures
- Supports human pluripotent stem cell attachment and survival better than wild type vitronectin
- Optimal environment for feeder-free pluripotent stem cell culture
- Defined, recombinant human protein that reduces variability in your PSC cultures compared to human-plasma derived vitronectin and standard basement membrane extracts (BMEs)
Specifications
Specifications
| Concentration | 500 μg/mL |
| For Use With (Application) | Stem Cell Culture |
| Purification Method | HPLC |
| Quantity | 1 mL = 0.5 mg |
| Species | Human |
| Recombinant | Recombinant |
| Content And Storage | Store at -80°C and protect from light. |
| Classification | AOF |
| Endotoxin Level | Very Low |
| Shipping Condition | Dry ice |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.