missing translation for 'onlineSavingsMsg'
Learn More

Vitamin K1 98.0+%, TCI America™

Catalog No. p-7134710
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
P06425G 5 g
1 options

Catalog No. P06425G

Supplier: TCI America P06425G

Only null left

Chemical Identifiers

CAS 84-80-0
Molecular Formula C31H46O2
Molecular Weight (g/mol) 450.707
MDL Number MFCD00214063
InChI Key MBWXNTAXLNYFJB-NKFFZRIASA-N
Synonym vitamin k1, phytonadione, phylloquinone, phytylmenadione, phyllochinon, phytomenadione, alpha-phylloquinone, 3-phytylmenadione, konakion, 2-methyl-3-phytyl-1,4-naphthoquinone
PubChem CID 5284607
ChEBI CHEBI:18067
IUPAC Name 2-methyl-3-[(E,7R,11R)-3,7,11,15-tetramethylhexadec-2-enyl]naphthalene-1,4-dione
SMILES CC1=C(C(=O)C2=CC=CC=C2C1=O)CC=C(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

Melting Point -20°C
Quantity 5 g
Formula Weight 450.71
Percent Purity ≥98.0% (E)
Physical Form Liquid
Chemical Name or Material Vitamin K1
RTECSNumber QJ5800000
TSCA Yes
Recommended Storage Refrigerator