missing translation for 'onlineSavingsMsg'
Learn More

Vitamin E Acetate, USP, 96-102%, Spectrum™ Chemical

VI140, 7695-91-2, C31H52O3

$454.42 - $2425.27

Chemical Identifiers

CAS 7695-91-2
Molecular Formula C31H52O3
Molecular Weight (g/mol) 472.75
InChI Key ZAKOWWREFLAJOT-CEFNRUSXSA-N
IUPAC Name (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl acetate
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18607722
View Documents
Spectrum Chemical Mfg Cor
VI140100ML
100 mL Amber Glass Bottle
Each for $454.42
Only null left
 
18607723
View Documents
Spectrum Chemical Mfg Cor
VI1401LTGL
1 L Amber Glass Bottle
Each for $2,425.27
Only null left
 
Description

Description

Spectrum™ Chemical Vitamin E Acetate, USP is used as an antioxidant. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

7695-91-2
472.75
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl acetate
C31H52O3
ZAKOWWREFLAJOT-CEFNRUSXSA-N
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1

Specifications

1.0 mL
1
C31H52O3
ZAKOWWREFLAJOT-CEFNRUSXSA-N
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl acetate
96 to 102%
7695-91-2
Amber Glass Bottle
100 mL
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1
472.75
USP
Videos
SDS
Documents

Documents