missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitamin D3 Alexa Fluor 594-conjugated Antibody, R&D Systems™
Description
Vitamin D3 Monoclonal specifically detects Vitamin D3 in Multi-species samples. It is validated for Flow Cytometry (Intracellular Staining).
Specifications
Specifications
| Antigen | Vitamin D3 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 685503 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 594 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Host Species | Mouse |
| Immunogen | 25-Hydroxyvitamin D 3 |
| Show More |
For Research Use Only (RUO)