missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies Pipette Tips: Non-Sterile

For use with Ovation and MLA Brand pipetters

$143.59 - $295.76

Specifications

Filtered Non-filtered
Sterility Non-sterile
Color Clear
Rack Color Blue
Product Type Pipette Tips
View More Specs

Products 13
Catalog Number Mfr. No. Volume (Metric) Packaging Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Packaging Price Quantity  
21-341-20
View Documents
N/A 250 μL Econo-Pak: Plastic Container
Pack of 1000 Each for $246.07
Case of 10 Pack for $2,460.70
Only null left
 
21-343-20
View Documents
N/A 1250 μL Econo-Pak: Plastic Container
Pack of 750 Each for $197.14
Case of 10 Pack for $1,971.40
Only null left
 
22-146-630
View Documents
VistaLab™ Technologies
40581000
25 μL VistaBulk: Plastic Bag
Pack of 1000 Each for $143.59
Case of 10 Pack for $1,435.90
Only null left
 
22-146-636
View Documents
VistaLab™ Technologies
40582000
250 μL VistaBulk: Plastic Bag
Pack of 1000 Each for $148.97
Case of 10 Pack for $1,489.70
Only null left
 
22-146-643
View Documents
VistaLab™ Technologies
40583000
1250 μL VistaBulk: Plastic Bag
Pack of 1000 Each for $177.72
Case of 10 Pack for $1,777.20
Only null left
 
22-146-350
View Documents
N/A 25 μL VistaRak: 5 racks
Case of 960 Each for $245.55
Only null left
 
22-146-351
View Documents
VistaLab™ Technologies
40602004
250 μL VistaRak: 5 racks
Case of 960 Each for $216.69
Only null left
 
22-146-352
View Documents
N/A 1250 μL VistaRak: 5 racks
Case of 960 Each for $292.97
Only null left
 
22-146-353
View Documents
N/A 25 μL VistaStak: 3 layers
Pack of 576 Each for $169.66
Only null left
 
22-146-354
View Documents
N/A 250 μL VistaStak: 5 layers
Pack of 960 Each for $295.76
Case of 10 Pack for $2,957.60
Only null left
 
22-146-355
View Documents
N/A 1250 μL VistaStak: 3 layers
Pack of 576 Each for $162.10
Case of 10 Pack for $1,621.00
Only null left
 
21-341
View Documents
N/A 250 μL Stacked Rack: 5 layers
Pack of 1000 Each for $235.59
Case of 10 Pack for $2,355.90
Only null left
 
21-343
View Documents
N/A 1250 μL Stacked Rack: 3 layers
Pack of 600 Each for $177.86
Case of 10 Pack for $1,778.60
Only null left
 
Description

Description

  • Non-Sterile tips packaged in VistaRak, VistaStak, Stacked Rack, Bulk
  • VistaRak: 5 racks, 192 tips/rack, VistaStak: 3 or 5 Layers of 192 tips, Stacked Rack: 3 or 5 layers of 200 tips, Bulk: 750-1000 tips in a plastic bag or containers
  • Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
  • All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time
Specifications

Specifications

Non-filtered
Clear
Pipette Tips
No
Non-sterile
Blue
Single-use
Polypropylene
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'