missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Tanks
- Reservoirs and Liners
- VistaLab™ Technologies Reagent Reservoirs
VistaLab™ Technologies Reagent Reservoirs
Catalog No. 22145102

Change view
Click to view available options
Capacity (Metric):
100 mL (with lid)
25 mL (divided)
10 mL
25 mL
50 mL
100 mL
Sterility:
Non-sterile
Sterile
Packaging:
Bulk
Individually packed
Individually Wrapped
Quantity:
10/bag x 10; 100/Cs.
100/Cs.
20/bag x 10; 200/Cs.
200 in. 20 bags of 10
50/Cs.
24 product options available for selection
| Catalog No. | Capacity (Metric) | Sterility | Packaging | Quantity |
|---|---|---|---|---|
| 22145102 | 25 mL (divided) | Non-sterile | Bulk | 10/bag x 10; 100/Cs. |
| 2138127A | 100 mL | Sterile | Bulk | 10/bag x 10; 100/Cs. |
| 2138127B | 100 mL | Sterile | Individually Wrapped | 50/Cs. |
| 2138127E | 25 mL (divided) | Sterile | Bulk | 10/bag x 10; 100/Cs. |
| 21385111 | 100 mL (with lid) | Sterile | Individually Wrapped | 50/Cs. |
| 2138127D | 25 mL | Sterile | Individually Wrapped | 50/Cs. |
| 2138127G | 50 mL | Sterile | Bulk | 10/bag x 10; 100/Cs. |
| 2138127H | 50 mL | Sterile | Individually Wrapped | 50/Cs. |
| 21385101 | 100 mL | Sterile | Individually Wrapped | 100/Cs. |
| 21385100 | 100 mL | Sterile | Bulk | 20/bag x 10; 200/Cs. |
| 21385104 | 50 mL | Sterile | Bulk | 20/bag x 10; 200/Cs. |
| 21385102 | 25 mL | Sterile | Bulk | 20/bag x 10; 200/Cs. |
| 21385105 | 50 mL | Sterile | Individually Wrapped | 100/Cs. |
| 21385103 | 25 mL | Sterile | Individually Wrapped | 100/Cs. |
| 22145101 | 25 mL | Non-sterile | Bulk | 10/bag x 10; 100/Cs. |
| 22145103 | 50 mL | Non-sterile | Bulk | 10/bag x 10; 100/Cs. |
| 22145100 | 100 mL | Non-sterile | Bulk | 10/bag x 10; 100/Cs. |
| 2138127C | 25 mL | Sterile | Bulk | 10/bag x 10; 100/Cs. |
| 2138127F | 25 mL (divided) | Sterile | Individually Wrapped | 50/Cs. |
| 21381090 | 10 mL | Sterile | Bulk | 100/Cs. |
| 21381091 | 10 mL | Sterile | Individually packed | 50/Cs. |
| 21381092 | 10 mL | Sterile | Bulk | 200 in. 20 bags of 10 |
| 21381093 | 10 mL | Sterile | Individually packed | 100/Cs. |
| 21381094 | 10 mL | Non-sterile | Bulk | 100/Cs. |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
Catalog No.
22145102
Supplier
VistaLab™ Technologies
Supplier No.
40541004
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
- Polystyrene with trimmed base corners
- Distinct pour-off spouts to empty reservoirs without spills
- Well-marked graduated increments on inside wall minimize reagent waste
- Wide base and sturdy construction provide stability and help to avoid spills
- Molded corner structures allow easy separation of bulk packaged reservoirs
- Available in 25, 50, 100mL and 1.8L volumes; 25mL available with divided configuation for multichannel pipetting of two reagents simultaneously
- 100mL model (Cat. No. 21-385-111) is supplied with lid
- Available individually wrapped or bulk packaged, gamma sterilized or nonsterile
Order Info
Reservoir lid sold separately (Cat. No. 21-385-110).
Specifications
Specifications
| Certifications/Compliance | Pyrogen-free |
| Color | White |
| Capacity (Metric) | 25 mL (divided) |
| Material | Polystyrene |
| Sterility | Non-sterile |
| Type | Reagent Reservoir |
| For Use With (Application) | General Liquid Handling |
| Height (English) | 7.5 in. |
| Length (English) | 13 in. |
| Packaging | Bulk |
| Quantity | 10/bag x 10; 100/Cs. |
| Width (English) | 7 in. |
| Autoclavable | Not Autoclavable |
| Channels | 1 |
| Disposable | Single-use |
| Bottom Shape | V-bottom |
| Show More Show Less |