missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies Reagent Reservoirs: 12-Channel

Description
- Polystyrene with trimmed base corners
- Distinct pour-off spouts to empty reservoirs without spills
- Well-marked graduated increments on inside wall minimize reagent waste
- Wide base and sturdy construction provide stability and help to avoid spills
- Molded corner structures allow easy separation of bulk packaged reservoirs
- 100mL model (Cat. No. 21-382-111) supplied with lid
- Available individually wrapped or bulk packaged, gamma sterilized or nonsterile
- 5 mL capacity / well
Specifications
Specifications
| Certifications/Compliance | RNase/DNase, Pyrogen-free |
| Color | Clear |
| Capacity (Metric) | 60 mL |
| Material | Polystyrene |
| Sterility | Non-sterile |
| Type | Reagent Reservoir |
| For Use With (Application) | General Liquid Handling |
| Height (English) | 6 in. |
| Includes | Lid |
| Length (English) | 11 in. |
| Show More |