missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies Reagent Reservoirs: 12-Channel

Unique trough-within-trough design maximizes reagent recovery.

Supplier:  VistaLab™ Technologies 40541010

 View more versions of this product

Catalog No. 21-385-122


Only null left
Explore available promotions

Description

Description

  • Polystyrene with trimmed base corners
  • Distinct pour-off spouts to empty reservoirs without spills
  • Well-marked graduated increments on inside wall minimize reagent waste
  • Wide base and sturdy construction provide stability and help to avoid spills
  • Molded corner structures allow easy separation of bulk packaged reservoirs
  • 100mL model (Cat. No. 21-382-111) supplied with lid
  • Available individually wrapped or bulk packaged, gamma sterilized or nonsterile
  • 5 mL capacity / well
Specifications

Specifications

RNase/DNase, Pyrogen-free
60 mL
Non-sterile
General Liquid Handling
Lid
Bulk
5 mL
Autoclavable
Single-use
Clear
Polypropylene
Reagent Reservoir
5 in.
12 in.
10/Cs.
6 in.
12
V-bottom
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions