missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies Reagent Reservoirs: 12-Channel
Unique trough-within-trough design maximizes reagent recovery.
Supplier: VistaLab™ Technologies 30541011
Description
- Polystyrene with trimmed base corners
- Distinct pour-off spouts to empty reservoirs without spills
- Well-marked graduated increments on inside wall minimize reagent waste
- Wide base and sturdy construction provide stability and help to avoid spills
- Molded corner structures allow easy separation of bulk packaged reservoirs
- 100mL model (Cat. No. 21-382-111) supplied with lid
- Available individually wrapped or bulk packaged, gamma sterilized or nonsterile
- 5 mL capacity / well
Specifications
| RNase/DNase, Pyrogen-free | |
| 60 mL | |
| Sterile | |
| General Liquid Handling | |
| Lid | |
| Individually Wrapped | |
| 5 mL | |
| Autoclavable | |
| Single-use |
| Clear | |
| Polypropylene | |
| Reagent Reservoir | |
| 8 in. | |
| 12 in. | |
| 25/Cs. | |
| 8 in. | |
| 12 | |
| V-bottom |