missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VISTA Recombinant Armenian Hamster Monoclonal Antibody (MH5A)

Armenian Hamster Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551819
Description
This antibody specifically binds PD-1H.
VISTA (V-domain Ig containing suppressor of T-cell activation), also known as B7-H5 or PD-1H, is a product of the VSIR gene. Although VISTA bears sequence homology to PD-L1 it has a different expression pattern. In both mouse and humans, VISTA is expressed predominantly on hematopoietic cells with the highest densities on myeloid cells, and lower levels on T cells. It is usually absent on B cells. VISTA expressed by antigen presenting cells has been shown to directly suppress proliferation and cytokine production of CD4 and CD8 T cells, making it a target for cancer immunotherapy. The reported ligands of VISTA include CD28H, VSIG-3 and VSIG-8.Chemical Identifiers
| VISTA | |
| 1 mg/mL | |
| ELISA, Functional Assay, Inhibition Assays, Immunohistochemistry | |
| Unconjugated | |
| Armenian Hamster | |
| RUO | |
| PBS with 0.02% ProClin 300 | |
| Q9D659 | |
| 74048 | |
| The antibody was generated by immunising Armenian hamsters with mPD-1HIg. | |
| Antibody |
| MH5A | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG λ | |
| Mouse | |
| VSIR | |
| 4632428N05Rik; B7H5; B7-H5; C10orf54; C1H10orf54; C28H10orf54; C4H10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; VSIR; V-type immunoglobulin domain-containing suppressor of T-cell activation | |
| VSIR | |
| Primary | |
| Protein A |
Specifications
| VISTA | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Q9D659 | |
| VSIR | |
| The antibody was generated by immunising Armenian hamsters with mPD-1HIg. | |
| 200 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG λ |
| ELISA, Functional Assay, Inhibition Assays, Immunohistochemistry | |
| MH5A | |
| Unconjugated | |
| VSIR | |
| 4632428N05Rik; B7H5; B7-H5; C10orf54; C1H10orf54; C28H10orf54; C4H10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; VSIR; V-type immunoglobulin domain-containing suppressor of T-cell activation | |
| Armenian Hamster | |
| Protein A | |
| RUO | |
| 74048 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |