missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VISTA Monoclonal Antibody (B7H5DS8), APC, eBioscience™, Invitrogen™

Catalog No. 17108841
Encompass
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17108841 25 Tests
17108842 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17108841 Supplier Invitrogen™ Supplier No. 17108841
Only null left

Mouse Monoclonal Antibody

Description: This B7H5DS8 monoclonal antibody recognizes human VISTA also known as B7-H5 or PD-H1. This clone has been tested and shown not to cross-react with human B7-H4, B7-H5 and B7-H7. This B7H5DS8 antibody will work in flow cytometry only on live and not paraformaldehyde-fixed cells. Applications Reported: This B7H5DS8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This B7H5DS8 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser

VISTA (V-domain Ig containing suppressor of T-cell activation), also known as B7-H5 or PD-1H, is a product of the VSIR gene. Although VISTA bears sequence homology to PD-L1 it has a different expression pattern. In both mouse and humans, VISTA is expressed predominantly on hematopoietic cells with the highest densities on myeloid cells, and lower levels on T cells. It is usually absent on B cells. VISTA expressed by antigen presenting cells has been shown to directly suppress proliferation and cytokine production of CD4 and CD8 T cells, making it a target for cancer immunotherapy. The reported ligands of VISTA include CD28H, VSIG-3 and VSIG-8.

Specifications

Antigen VISTA
Applications Flow Cytometry
Classification Monoclonal
Clone B7H5DS8
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene VSIR
Gene Accession No. Q9H7M9
Gene Alias 4632428N05Rik; B7H5; B7-H5; C10orf54; C1H10orf54; C28H10orf54; C4H10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; VSIR; V-type immunoglobulin domain-containing suppressor of T-cell activation
Gene Symbols VSIR
Host Species Mouse
Immunogen VISTA-Fc protein.
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64115
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less