missing translation for 'onlineSavingsMsg'
Learn More

Koehler™ Instrument Viscosity Reference Standards

For calibration and verification of kinematic and dynamic viscosity test equipment, both manual and automatic

Supplier:  Koehler™ Instrument 355002600

 View more versions of this product

Catalog No. 13-617-66


Only null left

Description

Description

For calibration and verification of kinematic and dynamic viscosity test equipment, both manual and automatic

• Standards are based upon the NIST value of 1.0034 cSt for water at 20°C (68°F)
• Traceable to National Standards; manufactured according to ASTM D2162 specifications
• Supplied with ISO/IEC 17025 Certification Report
• Fully compliant with ASTM and related test procedures
• Calibrated to a precision of ±0.2% for viscosity and kinematic viscosity
• Each standard is calibrated at a minimum of five temperatures
• Supplied in 500mL amber bottle, with full certification and MSDS

Specifications

Specifications

518 cSt at 40°C
Calibration and Verification of Kinematic and Dynamic Viscosity Test
Kinematic and Dynamic Viscosity Test Equipment
500 mL
Liquid
ISO/IEC 17025, ASTM D 2162, NIST
Viscosity Standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions