missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mirus Bio VirusGEN™ AAV Transfection Kit
Description
- TransIT-VirusGEN Transfection Reagent is designed to enhance delivery of packaging and transfer plasmids to adherent and suspension HEK 293 cell types to increase recombinant adeno-associated virus (AAV) and lentivirus production.
- Kit includes: TransIT-VirusGEN Transfection Reagent, VirusGEN AAV Complex formation Solution and Enhancer
- TransIT-VirusGEN™ is ideal for academic research and biopharmaceutical discovery. Titers can be further increased with optimized enhancers included in the VirusGEN AAV Transfection Kit and VirusGEN LV Transfection Kit.
- Key benefits:
- Performance: Efficient DNA delivery for production of high-titer viral vectors
- Scalability: Efficient production from less than 1 mL to large scale culturesFlexibility: Compatible with different recombinant virus and cell culture systems
- Animal Origin Free: Fully synthetic transfection reagent formulation
Specifications
Specifications
| Cell Type | 293-derived Cells |
| Format | Bottle |
| Promoter | None (Promoterless) |
| Transfection Technique | Chemical Transfection |
| Product Type | AAV Transfection Kit |
| Volume (Metric) | 150 mL |