missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ViralSEQ™ Vesivirus Detection Kit
GREENER_CHOICE

Catalog No. 4448398C
Change view
Click to view available options
Quantity:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4448398C 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4448398C Supplier Applied Biosystems™ Supplier No. 4448398C

The ViralSEQ Vesivirus Detection Assay is a PCR-based test that enables users to quickly isolate and detect Vesivirus 2117 and all other known strains with high sensitivity and specificity.

Viral contamination in mammalian product manufacturing presents a serious risk to the manufacturing process, the manufacturing facility, and the integrity of the product. Vesivirus 2117 is an RNA virus that has re-emerged as a potential threat to mammalian cell culture production. To meet the challenge of rapid, specific, and sensitive detection of Vesivirus, Applied Biosystems has developed the first commercially available integrated kit for isolation and detection of Vesivirus from mammalian cell culture samples.

The ViralSEQ Vesivirus Detection Assay is a PCR-based test that enables users to quickly isolate and detect Vesivirus 2117 and all other known strains with high sensitivity and specificity. The test utilizes PrepSEQ sample preparation magnetic beads to isolate viral RNA from a wide range of sample types, combined with TaqMan-based technology for sensitive and specific detection of the viral RNA. The kit also includes a proprietary positive control template that helps eliminate the possibility of false positive results arising from inadvertent contamination of test samples with the positive control.

  • Detection of known Vesivirus strains
  • Rapid time-to-results in less than 5 hours
  • TaqMan-based real-time PCR assay
  • Demonstrated sensitivity and specificity
  • Proprietary optimized sample preparation for high RNA recovery

Exclusion Panel

The ViralSEQ Assay has been demonstrated to be specific for Vesivirus and does not detect RNA purified from related and unrelated species and organisms.

  • Bacillus: 2.5 ng
  • Chicken: 5 ng
  • Clostridium: 2.5 ng
  • Dog: 5 ng
  • E. coli: 2.5 ng
  • Human: 5 ng
  • Lactoccus: 2.5 ng
  • MDCK: 5 NG
  • Shigella: 2.5 ng
  • Rat: 5 ng
  • Yeast: 5 ng
  • CHO: 500 ng
  • Human RNA: 1 μg

Specifications

Content And Storage Contains RT-PCR assay, buffer and enzyme mix, positive control and negative control
Detection Method Primer-probe
Sample Type RNA
For Use With (Application) 1-Step qRT-PCR
No. of Reactions 100 Reactions
Product Line ViralSEQ
Product Type Vesivirus Detection Kit
Quantity 100 reactions
Shipping Condition Dry Ice
Target Viral RNA

For Research Use Only. Not for use in diagnostic procedures.