missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Viral Production Cells 2.0
Description
Gibco Viral Production Cells 2.0 are a clonal cell line derived from the HEK293F parental cell line and a core component of the AAV-MAX Helper-Free AAV Production System.
- Adapted to the chemically defined Viral Production Medium and maintained in suspension culture in that medium
- Highly transfectable using the AAV-MAX Transfection Kit for adeno-associated virus (AAV) production
- Supplied frozen in a vial containing 1 mL of cells at 1 x 107 viable cells/mL in Viral Production Medium and 10% DMSO
- Should be thawed directly into Viral Production Medium supplemented with 4 mM GlutaMAX Supplement (Cat. No. 35050038) for optimal performance
- Clonal, 293F-derived, high-producer cell line
- Absence of SV40 large T antigen
- Optimized for high-density, suspension culture (>12 million cells/mL) in chemically defined medium
- Enable production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
- Robust scalability and passage stability
Specifications
Specifications
| Cell State | Frozen |
| Concentration | 1 x 107 cells/mL |
| Content And Storage | 1 cryovial, store in vapor phase liquid nitrogen |
| Cell Line | Viral Production Cells 2.0 |
| Cell Type | Kidney (Embryonic) |
| Culture Type | Suspension Cell Culture |
| Form | Frozen |
| Species | Human |
| Format | 1 Vial |
| For Use With (Application) | AAV Production |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.