missing translation for 'onlineSavingsMsg'
Learn More

Vinyltris(2-methoxyethoxy)silane 96.0+%, TCI America™

Catalog No. V0048500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0048500ML 500 mL
V004825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0048500ML Supplier TCI America Supplier No. V0048500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1067-53-4
Molecular Formula C11H24O6Si
Molecular Weight (g/mol) 280.392
MDL Number MFCD00008500
InChI Key WOXXJEVNDJOOLV-UHFFFAOYSA-N
Synonym tris 2-methoxyethoxy vinylsilane, vinyltris 2-methoxyethoxy silane, vinyl tris 2-methoxyethoxy silane, silicone a-172, 6-2-methoxyethoxy-6-vinyl-2,5,7,10-tetraoxa-6-silaundecane, tris methoxyethxy vinylsilane, tris methoxyethoxy vinylsilane, vinyltris methoxyethoxy silane, nuca 172, 2,5,7,10-tetraoxa-6-silaundecane, 6-ethenyl-6-2-methoxyethoxy
PubChem CID 14025
IUPAC Name ethenyl-tris(2-methoxyethoxy)silane
SMILES COCCO[Si](C=C)(OCCOC)OCCOC

Specifications

Color Colorless
Boiling Point 113°C
Quantity 500 mL
Formula Weight 280.39
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyltris(2-methoxyethoxy)silane
EINECSNumber (2)-2067
RTECSNumber VV6826000
TSCA Yes