accessibility menu, dialog, popup

UserName

H3COOSiOOCH3OOCH3CH2

CAS RN 1067-53-4

IUPAC Name: 6-ethenyl-6-(2-methoxyethoxy)-2,5,7,10-tetraoxa-6-silaundecane
Synonyms: 6-ethenyl-6-(2-methoxyethoxy)-2,5,7,10-tetraoxa-6-silaundecane
Molecular Weight (g/mol): 280.39
Molecular Formula: C11H24O6Si
InChi Key: WOXXJEVNDJOOLV-UHFFFAOYSA-N
SMILES: COCCO[Si](OCCOC)(OCCOC)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results