Learn More
Vinyltriphenylphosphonium bromide, 97%
CAS: 5044-52-0 | C20H18BrP | 369.24 g/mol
$1546.41 - $1546.41
Chemical Identifiers
| CAS | 5044-52-0 |
|---|---|
| Molecular Formula | C20H18BrP |
| Molecular Weight (g/mol) | 369.24 |
| MDL Number | MFCD00011807 |
| InChI Key | VRAYVWUMBAJVGH-UHFFFAOYSA-M |
| Synonym | vinyltriphenylphosphonium bromide, triphenyl vinyl phosphonium bromide, schweizer's reagent, ethenyltriphenylphosphonium bromide, ethenyl triphenyl phosphanium bromide, ethenyltriphenylphosphanium bromide, triphenyl-vinyl-phosphonium; bromide, pubchem16334, acmc-1aw2n, vinyltriphenylphosphine bromide |
| PubChem CID | 2723893 |
| IUPAC Name | ethenyl(triphenyl)phosphanium;bromide |
| SMILES | [Br-].C=C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0070822
|
Thermo Scientific Chemicals
L0070822 |
100 g |
Each for $1,546.41
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5044-52-0 | |
| 369.24 | |
| VRAYVWUMBAJVGH-UHFFFAOYSA-M | |
| 2723893 | |
| [Br-].C=C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C20H18BrP | |
| MFCD00011807 | |
| vinyltriphenylphosphonium bromide, triphenyl vinyl phosphonium bromide, schweizer's reagent, ethenyltriphenylphosphonium bromide, ethenyl triphenyl phosphanium bromide, ethenyltriphenylphosphanium bromide, triphenyl-vinyl-phosphonium; bromide, pubchem16334, acmc-1aw2n, vinyltriphenylphosphine bromide | |
| ethenyl(triphenyl)phosphanium;bromide |
Specifications
| 5044-52-0 | |
| C20H18BrP | |
| 100 g | |
| Hygroscopic; Light sensitive | |
| VRAYVWUMBAJVGH-UHFFFAOYSA-M | |
| ethenyl(triphenyl)phosphanium;bromide | |
| 2723893 | |
| 97% |
| 182°C to 186°C | |
| MFCD00011807 | |
| 4168860 | |
| vinyltriphenylphosphonium bromide, triphenyl vinyl phosphonium bromide, schweizer's reagent, ethenyltriphenylphosphonium bromide, ethenyl triphenyl phosphanium bromide, ethenyltriphenylphosphanium bromide, triphenyl-vinyl-phosphonium; bromide, pubchem16334, acmc-1aw2n, vinyltriphenylphosphine bromide | |
| [Br-].C=C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 369.24 | |
| 369.25 | |
| Vinyltriphenylphosphonium bromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 225-740-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only