missing translation for 'onlineSavingsMsg'
Learn More

Vinyltriphenylphosphonium bromide, 97%

Catalog No. AAL0070822
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0070822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0070822 Supplier Thermo Scientific Chemicals Supplier No. L0070822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5044-52-0 | C20H18BrP | 369.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5044-52-0
Molecular Formula C20H18BrP
Molecular Weight (g/mol) 369.24
MDL Number MFCD00011807
InChI Key VRAYVWUMBAJVGH-UHFFFAOYSA-M
Synonym vinyltriphenylphosphonium bromide, triphenyl vinyl phosphonium bromide, schweizer's reagent, ethenyltriphenylphosphonium bromide, ethenyl triphenyl phosphanium bromide, ethenyltriphenylphosphanium bromide, triphenyl-vinyl-phosphonium; bromide, pubchem16334, acmc-1aw2n, vinyltriphenylphosphine bromide
PubChem CID 2723893
IUPAC Name ethenyl(triphenyl)phosphanium;bromide
SMILES [Br-].C=C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 182°C to 186°C
Quantity 100 g
Beilstein 4168860
Sensitivity Hygroscopic; Light sensitive
Formula Weight 369.25
Percent Purity 97%
Chemical Name or Material Vinyltriphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 225-740-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only