missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinyltoluene Monomer (m- and p- mixture) (stabilized with TBC) 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | -77°C |
| Color | Colorless |
| Boiling Point | 173°C |
| Quantity | 500 mL |
| UN Number | 2618 |
| Formula Weight | 118.18 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Vinyltoluene Monomer (m- and p- mixture) (stabilized with TBC) |
Safety and Handling
| EINECSNumber | (3)-0008 |
| RTECSNumber | WL5075000 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |