missing translation for 'onlineSavingsMsg'
Learn More

Vinyltoluene Monomer (m- and p- mixture) (stabilized with TBC) 98.0+%, TCI America™

Catalog No. V002825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V002825ML 25 mL
V0028500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V002825ML Supplier TCI America Supplier No. V002825ML
Only null left

Chemical Identifiers

CAS 25013-15-4
Molecular Formula C9H10
MDL Number MFCD00168066
Synonym Methylstyrene Monomer

Specifications

Melting Point -77°C
Color Colorless
Boiling Point 173°C
Quantity 25 mL
UN Number 2618
Formula Weight 118.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyltoluene Monomer (m- and p- mixture) (stabilized with TBC)
EINECSNumber (3)-0008
RTECSNumber WL5075000
TSCA Yes
Recommended Storage Refrigerator