missing translation for 'onlineSavingsMsg'
Learn More

Vinylsulfonic Acid 97.0+%, TCI America™

Catalog No. V01341G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V01341G 1 g
V01345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V01341G Supplier TCI America Supplier No. V01341G
Only null left

Chemical Identifiers

CAS 1184-84-5
Molecular Formula C2H3NaO3S
Molecular Weight (g/mol) 130.09
MDL Number MFCD09743544
InChI Key BWYYYTVSBPRQCN-UHFFFAOYSA-M
PubChem CID 62474
IUPAC Name sodium ethenesulfonate
SMILES [Na+].[O-]S(=O)(=O)C=C

Specifications

pH 0.97 (0.1mol 22.3°C)
Boiling Point 115°C
Quantity 1 g
UN Number 3265
Formula Weight 108.11
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Vinylsulfonic Acid
TSCA No
Recommended Storage Refrigerator