accessibility menu, dialog, popup

UserName

CAS RN 1184-84-5

NaOSOOH2C

CAS RN 1184-84-5

IUPAC Name: sodium ethenesulfonate
Synonyms: sodium apolate 阿朴酸钠
Molecular Weight (g/mol): 130.09
Molecular Formula: C2H3NaO3S
InChi Key: BWYYYTVSBPRQCN-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results