missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinylene Carbonate (stabilized with BHT) 98.0+%, TCI America™
Supplier: TCI America V00155G
Specifications
| Vinylene Carbonate (stabilized with BHT) | |
| White-Yellow | |
| C3H2O3 | |
| 5 g | |
| VAYTZRYEBVHVLE-UHFFFAOYSA-N | |
| 2H-1,3-dioxol-2-one | |
| 13385 | |
| ≥98.0% (GC) |
| 872-36-6 | |
| 60°C | |
| MFCD00005380 | |
| vinylene carbonate, vinyl carbonate, carbonic acid, cyclic vinylene ester, wln: t5ovoj, vinylenecarbonate, vinyleny carbonate, carbonic acid vinylene, 1,3-dioxo-2-one, vinylene carbonate vc, acmc-209qj3 | |
| O=C1OC=CO1 | |
| 86.05 | |
| 86.05 | |
| Crystalline Lumps |
Chemical Identifiers
| 872-36-6 | |
| 86.05 | |
| VAYTZRYEBVHVLE-UHFFFAOYSA-N | |
| 13385 | |
| O=C1OC=CO1 |
| C3H2O3 | |
| MFCD00005380 | |
| vinylene carbonate, vinyl carbonate, carbonic acid, cyclic vinylene ester, wln: t5ovoj, vinylenecarbonate, vinyleny carbonate, carbonic acid vinylene, 1,3-dioxo-2-one, vinylene carbonate vc, acmc-209qj3 | |
| 2H-1,3-dioxol-2-one |
Safety and Handling
EINECSNumber : (5)-6636&(5)-6744
RTECSNumber : FG3325000
TSCA : Yes
Recommended Storage : Refrigerator