missing translation for 'onlineSavingsMsg'
Learn More

Vinylene Carbonate (stabilized with BHT) 98.0+%, TCI America™

Catalog No. p-7137888
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00155G 5 g
V001525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V00155G Supplier TCI America Supplier No. V00155G
Only null left

Chemical Identifiers

CAS 872-36-6
Molecular Formula C3H2O3
Molecular Weight (g/mol) 86.05
MDL Number MFCD00005380
InChI Key VAYTZRYEBVHVLE-UHFFFAOYSA-N
Synonym vinylene carbonate, vinyl carbonate, carbonic acid, cyclic vinylene ester, wln: t5ovoj, vinylenecarbonate, vinyleny carbonate, carbonic acid vinylene, 1,3-dioxo-2-one, vinylene carbonate vc, acmc-209qj3
PubChem CID 13385
IUPAC Name 2H-1,3-dioxol-2-one
SMILES O=C1OC=CO1

Specifications

Color White-Yellow
Boiling Point 60°C
Quantity 5 g
Formula Weight 86.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Vinylene Carbonate (stabilized with BHT)
EINECSNumber (5)-6636&(5)-6744
RTECSNumber FG3325000
TSCA Yes
Recommended Storage Refrigerator