missing translation for 'onlineSavingsMsg'
Learn More

Vinylboronic anhydride pyridine complex, 95%

Catalog No. AAH2870303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2870303 1 g
AAH2870306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2870303 Supplier Thermo Scientific Chemicals Supplier No. H2870303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 95010-17-6 | C11H14B3NO3 | 240.667 g/mol

Reagent used for Suzuki-Miyaura cross-coupling, stereoselective synthesis via Palladium-catalyzed carboamination, Alkyl-connected 2-amino-6-vinylpurine (AVP) cross linking agent to cytosine base in RNA, Kaiser oxime resin-derived palladacycle as a recoverable polymeric precatalyst in Suzuki-Miyaura cross-coupling reactions in aqueous media and Kinetic resolution of phosphoryl and sulfonyl esters of binaphthol derivatives via Pd-catalyzed alcoholysis of their vinyl ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent used for Suzuki-Miyaura cross-coupling, stereoselective synthesis via Palladium-catalyzed carboamination, Alkyl-connected 2-amino-6-vinylpurine (AVP) cross linking agent to cytosine base in RNA, Kaiser oxime resin-derived palladacycle as a recoverable polymeric precatalyst in Suzuki-Miyaura cross-coupling reactions in aqueous media and Kinetic resolution of phosphoryl and sulfonyl esters of binaphthol derivatives via Pd-catalyzed alcoholysis of their vinyl ethers.

Solubility
Soluble in methanol.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.

Chemical Identifiers

CAS 95010-17-6
Molecular Formula C11H14B3NO3
Molecular Weight (g/mol) 240.667
MDL Number MFCD03839940
InChI Key YLHJACXHRQQNQR-UHFFFAOYSA-N
Synonym 2,4,6-trivinyl-1,3,5,2,4,6-trioxatriborinane compound with pyridine 1:1, trivinylboroxin pyridine complex, vinylboronic anhydride pyridine complex, 2,4,6-trivinylcyclotriboroxane-pyridine, 2,4,6-trivinylcyclotriboroxane pyridine, vinylboronic anhydride pyridine, 2,4,6-trivinylcyclotriboroxane pyridine complex, 2,4,6-trivinylcyclotriboroxane-pyridine complex, boroxin, triethenyl-, compd. with pyridine 1:1, pyridine; triethenyl-1,3,5,2,4,6-trioxatriborinane
PubChem CID 2734806
IUPAC Name pyridine;2,4,6-tris(ethenyl)-1,3,5,2,4,6-trioxatriborinane
SMILES B1(OB(OB(O1)C=C)C=C)C=C.C1=CC=NC=C1

Specifications

Melting Point 44°C to 52°C
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Soluble in methanol.
Formula Weight 240.67
Percent Purity 95%
Chemical Name or Material Vinylboronic anhydride pyridine complex
Hazard Category H228-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 672-662-4
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only