missing translation for 'onlineSavingsMsg'
Learn More

Vinyl Trifluoroacetate (stabilized with TBC) 97.0+%, TCI America™

Catalog No. p-7136662
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T12165G Supplier TCI America Supplier No. T12165G

Chemical Identifiers

CAS 433-28-3
Molecular Formula C4H3F3O2
Molecular Weight (g/mol) 140.061
MDL Number MFCD00003561
InChI Key ZBGRMWIREQJHPK-UHFFFAOYSA-N
Synonym vinyl trifluoroacetate, vinyltrifluoroacetate, trifluoroacetic acid vinyl ester, pubchem13129, acmc-1ahnj, vinyl 2,2,2-trifluoroacetate, zbgrmwireqjhpk-uhfffaoysa
PubChem CID 79011
IUPAC Name ethenyl 2,2,2-trifluoroacetate
SMILES C=COC(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 42°C
Quantity 5 g
UN Number 2924
Formula Weight 140.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl Trifluoroacetate (stabilized with TBC)
TSCA No
Recommended Storage Refrigerator