accessibility menu, dialog, popup

UserName

CAS RN 433-28-3

IUPAC Name: ethenyl 2,2,2-trifluoroacetate
Synonyms: ethenyl 2,2,2-trifluoroacetate
Molecular Weight (g/mol): 140.06
Molecular Formula: C4H3F3O2
InChi Key: ZBGRMWIREQJHPK-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)OC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results