missing translation for 'onlineSavingsMsg'
Learn More

Vinyl Stearate (stabilized with MEHQ) 95.0+%, TCI America™

Catalog No. S024125G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S024125G 25 g
S0241100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S024125G Supplier TCI America Supplier No. S024125G

Chemical Identifiers

CAS 111-63-7
Molecular Formula C20H38O2
Molecular Weight (g/mol) 310.522
MDL Number MFCD00026667
InChI Key AFSIMBWBBOJPJG-UHFFFAOYSA-N
Synonym Stearic Acid Vinyl Ester
PubChem CID 66077
IUPAC Name ethenyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OC=C

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 310.52
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Vinyl Stearate (stabilized with MEHQ)
EINECSNumber (2)-3474
TSCA Yes