accessibility menu, dialog, popup

UserName

H3COOCH2

CAS RN 111-63-7

IUPAC Name: ethenyl octadecanoate
Synonyms: ethenyl octadecanoate
Molecular Weight (g/mol): 310.52
Molecular Formula: C20H38O2
InChi Key: AFSIMBWBBOJPJG-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OC=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results