missing translation for 'onlineSavingsMsg'
Learn More

Vinyl n-Octanoate (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. p-7124671
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C089625ML 25 mL
C0896500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C089625ML Supplier TCI America Supplier No. C089625ML

Chemical Identifiers

CAS 818-44-0
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00048915
InChI Key QBDADGJLZNIRFQ-UHFFFAOYSA-N
Synonym n-Octanoic Acid Vinyl Ester
PubChem CID 13164
IUPAC Name ethenyl octanoate
SMILES CCCCCCCC(=O)OC=C

Specifications

Color Colorless
Boiling Point 135°C
Quantity 25 mL
Formula Weight 170.25
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Vinyl n-Octanoate (stabilized with MEHQ)
EINECSNumber (2)-0761
RTECSNumber RH0789000
TSCA No