missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™
$89.48 - $762.40
Chemical Identifiers
| CAS | 4245-37-8 |
|---|---|
| Molecular Formula | C6H8O2 |
| Molecular Weight (g/mol) | 112.128 |
| MDL Number | MFCD00059329 |
| InChI Key | FFYWKOUKJFCBAM-UHFFFAOYSA-N |
| Synonym | vinyl methacrylate, methacrylic acid, vinyl ester, methacrylic acid vinyl ester, 2-methyl-2-propenoic acid, ethenyl ester, 2-propenoic acid, 2-methyl-, ethenyl ester, methacrylic acid, vinyl ester 8ci, vinylmethacrylate, vinyl 2-methylacrylate #, acmc-1cr11 |
| PubChem CID | 77921 |
| IUPAC Name | ethenyl 2-methylprop-2-enoate |
| SMILES | CC(=C)C(=O)OC=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M072025ML
|
TCI America
M072025ML |
25 mL |
Each for $89.48
|
|
|||||
|
M0720100ML
|
TCI America
M0720100ML |
100 mL |
Each for $266.56
|
|
|||||
|
M0720500ML
|
TCI America
M0720500ML |
500 mL |
Each for $762.40
|
|
|||||
Chemical Identifiers
| 4245-37-8 | |
| 112.128 | |
| FFYWKOUKJFCBAM-UHFFFAOYSA-N | |
| 77921 | |
| CC(=C)C(=O)OC=C |
| C6H8O2 | |
| MFCD00059329 | |
| vinyl methacrylate, methacrylic acid, vinyl ester, methacrylic acid vinyl ester, 2-methyl-2-propenoic acid, ethenyl ester, 2-propenoic acid, 2-methyl-, ethenyl ester, methacrylic acid, vinyl ester 8ci, vinylmethacrylate, vinyl 2-methylacrylate #, acmc-1cr11 | |
| ethenyl 2-methylprop-2-enoate |
Specifications
| 4245-37-8 | |
| 112°C | |
| MFCD00059329 | |
| 3272 | |
| FFYWKOUKJFCBAM-UHFFFAOYSA-N | |
| ethenyl 2-methylprop-2-enoate | |
| 77921 | |
| ≥98.0% (GC) | |
| Vinyl Methacrylate (stabilized with MEHQ) |
| Colorless | |
| C6H8O2 | |
| 25 mL | |
| vinyl methacrylate, methacrylic acid, vinyl ester, methacrylic acid vinyl ester, 2-methyl-2-propenoic acid, ethenyl ester, 2-propenoic acid, 2-methyl-, ethenyl ester, methacrylic acid, vinyl ester 8ci, vinylmethacrylate, vinyl 2-methylacrylate #, acmc-1cr11 | |
| CC(=C)C(=O)OC=C | |
| 112.128 | |
| 112.13 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-4157
TSCA : Yes