missing translation for 'onlineSavingsMsg'
Learn More

Vinyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™

Supplier:  TCI America M0720500ML

 View more versions of this product

Catalog No. M0720500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Vinyl Methacrylate (stabilized with MEHQ)
Colorless
C6H8O2
500 mL
vinyl methacrylate, methacrylic acid, vinyl ester, methacrylic acid vinyl ester, 2-methyl-2-propenoic acid, ethenyl ester, 2-propenoic acid, 2-methyl-, ethenyl ester, methacrylic acid, vinyl ester 8ci, vinylmethacrylate, vinyl 2-methylacrylate #, acmc-1cr11
CC(=C)C(=O)OC=C
112.128
112.13
Liquid
4245-37-8
112°C
MFCD00059329
3272
FFYWKOUKJFCBAM-UHFFFAOYSA-N
ethenyl 2-methylprop-2-enoate
77921
≥98.0% (GC)

Chemical Identifiers

4245-37-8
112.128
FFYWKOUKJFCBAM-UHFFFAOYSA-N
77921
CC(=C)C(=O)OC=C
C6H8O2
MFCD00059329
vinyl methacrylate, methacrylic acid, vinyl ester, methacrylic acid vinyl ester, 2-methyl-2-propenoic acid, ethenyl ester, 2-propenoic acid, 2-methyl-, ethenyl ester, methacrylic acid, vinyl ester 8ci, vinylmethacrylate, vinyl 2-methylacrylate #, acmc-1cr11
ethenyl 2-methylprop-2-enoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-4157

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions